Assay Details
| Gene Symbol: Kpna1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008465.5 | NP_032491.2 | 11 - 12 | 1276 | 88 | |
| GenBank mRNA | AK028259.1 | - | 11 - 12 | 1277 | 88 | |
| AK028307.1 | - | 11 - 12 | 1295 | 88 | ||
| AK154800.1 | - | 11 - 12 | 1294 | 88 | ||
| BC006771.1 | - | 11 - 12 | 1261 | 3588534 | 88 | |
| U20619.1 | - | 11 - 12 | 1281 | 88 | ||
| U34228.1 | - | 10 - 11 | 1125 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kpna1
Genomic Map