Assay Details
| Gene Symbol: Kpna3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008466.5 | NP_032492.1 | 11 - 12 | 1055 | 58 | |
| GenBank mRNA | AF020772.1 | - | 11 - 12 | 899 | 58 | |
| AK011559.1 | - | 11 - 12 | 926 | 58 | ||
| AK049373.1 | - | 11 - 12 | 926 | 58 | ||
| AK161664.1 | - | 11 - 12 | 985 | 58 | ||
| BC026885.1 | - | 11 - 12 | 949 | 3979788 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kpna3
Genomic Map