Assay Details
| Gene Symbol: Kpna3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008466.5 | NP_032492.1 | 15 - 16 | 1528 | 83 | |
| GenBank mRNA | AF020772.1 | - | 15 - 16 | 1372 | 83 | |
| AK049373.1 | - | 15 - 16 | 1399 | 83 | ||
| BC026885.1 | - | 15 - 16 | 1422 | 3979788 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kpna3
Genomic Map