Assay Details
| Gene Symbol: Ltk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008523.2 | NP_032549.2 | 12 - 13 | 1530 | 59 | |
| NM_203345.2 | NP_976220.2 | 18 - 19 | 2376 | 59 | ||
| NM_206941.1 | NP_996824.1 | 13 - 14 | 1713 | 59 | ||
| NM_206942.2 | NP_996825.2 | 17 - 18 | 2193 | 59 | ||
| GenBank mRNA | BC047138.1 | - | 12 - 13 | 1530 | 6389834 | 59 |
| M90470.1 | - | 18 - 19 | 2376 | 59 | ||
| X07984.1 | - | 12 - 13 | 1504 | 59 | ||
| X52621.1 | - | 12 - 13 | 1519 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ltk
Genomic Map