Assay Details
| Gene Symbol: Met | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008591.2 | NP_032617.2 | 14 - 15 | 3241 | 66 | |
| GenBank mRNA | BC117969.1 | - | 12 - 13 | 2885 | 66 | |
| BC117970.1 | - | 12 - 13 | 2881 | 66 | ||
| CK332973.1 | - | 3 - 4 | 357 | 66 | ||
| Y00671.1 | - | 12 - 13 | 2886 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Met
Genomic Map