Assay Details
| Gene Symbol: Nnt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008710.3 | NP_032736.2 | 2951 | 88 | ||
| GenBank mRNA | AK033282.1 | - | 13 - 14 | 2139 | 88 | |
| AK040460.1 | - | 11 - 12 | 1817 | 88 | ||
| AK050478.1 | - | 12 - 13 | 2856 | 88 | ||
| AK052234.1 | - | 13 - 14 | 2183 | 88 | ||
| AK054332.1 | - | 13 - 14 | 2184 | 88 | ||
| AK083413.1 | - | 13 - 14 | 2182 | 88 | ||
| AK084546.1 | - | 13 - 14 | 2181 | 88 | ||
| AK084604.1 | - | 13 - 14 | 2181 | 88 | ||
| AK085903.1 | - | 11 - 12 | 1842 | 88 | ||
| AK088044.1 | - | 13 - 14 | 2933 | 88 | ||
| AK090115.1 | - | 2954 | 88 | |||
| AK129006.1 | - | 13 - 14 | 2928 | 88 | ||
| AK159694.1 | - | 15 - 16 | 2937 | 88 | ||
| AK168736.1 | - | 12 - 13 | 2734 | 88 | ||
| BC008518.1 | - | 13 - 14 | 2914 | 3601702 | 88 | |
| Z49204.1 | - | 12 - 13 | 2858 | 88 | ||
More Information
Set Membership: |
|

Nnt
Genomic Map