Assay Details
| Gene Symbol: Npr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008727.5 | NP_032753.5 | 12 - 13 | 2289 | 62 | |
| GenBank mRNA | AK135008.1 | - | 3 - 4 | 232 | 62 | |
| BC110659.1 | - | 12 - 13 | 2169 | 30847317 | 62 | |
| BC139767.1 | - | 12 - 13 | 2148 | 62 | ||
| L31932.1 | - | 12 - 13 | 1937 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Npr1
Genomic Map