Assay Details
| Gene Symbol: Npr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008727.5 | NP_032753.5 | 17 - 18 | 3043 | 84 | |
| GenBank mRNA | AK135008.1 | - | 8 - 9 | 986 | 84 | |
| BC110659.1 | - | 17 - 18 | 2923 | 30847317 | 84 | |
| BC139767.1 | - | 17 - 18 | 2902 | 84 | ||
| J05504.1 | - | 18 - 19 | 3027 | 84 | ||
| L31932.1 | - | 17 - 18 | 2691 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Npr1
Genomic Map