Assay Details
| Gene Symbol: Slc11a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008732.2 | NP_032758.2 | 16 - 17 | 1755 | 90 | |
| GenBank mRNA | BC019137.1 | - | 16 - 17 | 1729 | 5040976 | 90 |
| L33415.1 | - | 16 - 17 | 1683 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc11a2
Genomic Map