Assay Details
| Gene Symbol: Nrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008737.2 | NP_032763.2 | 2 - 3 | 729 | 86 | |
| GenBank mRNA | AK082992.1 | - | 2 - 3 | 782 | 86 | |
| AK135428.1 | - | 2 - 3 | 406 | 86 | ||
| AK141964.1 | - | 2 - 3 | 521 | 86 | ||
| AK148372.1 | - | 2 - 3 | 705 | 86 | ||
| AK159219.1 | - | 2 - 3 | 520 | 86 | ||
| AK164378.1 | - | 2 - 3 | 731 | 86 | ||
| BC006184.2 | - | 904 | 86 | |||
| BC026730.1 | - | 2 - 3 | 708 | 86 | ||
| BC032048.1 | - | 2 - 3 | 708 | 86 | ||
| BC060129.1 | - | 2 - 3 | 509 | 6409596 | 86 | |
| D50086.1 | - | 2 - 3 | 589 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nrp1
Genomic Map