Assay Details
| Gene Symbol: Nrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008737.2 | NP_032763.2 | 3 - 4 | 915 | 74 | |
| GenBank mRNA | AK082992.1 | - | 3 - 4 | 968 | 74 | |
| AK135428.1 | - | 3 - 4 | 592 | 74 | ||
| AK141964.1 | - | 3 - 4 | 707 | 74 | ||
| AK148372.1 | - | 3 - 4 | 891 | 74 | ||
| AK159219.1 | - | 3 - 4 | 706 | 74 | ||
| AK164378.1 | - | 3 - 4 | 917 | 74 | ||
| BC060129.1 | - | 3 - 4 | 695 | 6409596 | 74 | |
| D50086.1 | - | 3 - 4 | 775 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nrp1
Genomic Map