Assay Details
| Gene Symbol: Nrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008737.2 | NP_032763.2 | 12 - 13 | 2408 | 86 | |
| GenBank mRNA | AK141964.1 | - | 12 - 13 | 2200 | 86 | |
| AK148372.1 | - | 12 - 13 | 2384 | 86 | ||
| AK188315.1 | - | 3 - 4 | 264 | 86 | ||
| AK192303.1 | - | 3 - 4 | 209 | 86 | ||
| BC060129.1 | - | 12 - 13 | 2188 | 6409596 | 86 | |
| D50086.1 | - | 12 - 13 | 2268 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nrp1
Genomic Map