Assay Details
| Gene Symbol: Nsf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008740.4 | NP_032766.2 | 4 - 5 | 309 | 72 | |
| GenBank mRNA | AK042341.1 | - | 5 - 6 | 327 | 72 | |
| AK049281.1 | - | 4 - 5 | 279 | 72 | ||
| AK051430.1 | - | 4 - 5 | 278 | 72 | ||
| AK085086.1 | - | 4 - 5 | 263 | 72 | ||
| AK141986.1 | - | 4 - 5 | 278 | 72 | ||
| AK153905.1 | - | 4 - 5 | 278 | 72 | ||
| BC006627.1 | - | 4 - 5 | 314 | 3968733 | 72 | |
| BC019167.1 | - | 4 - 5 | 262 | 4506351 | 72 | |
| U10120.1 | - | 4 - 5 | 267 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nsf
Genomic Map