Assay Details
| Gene Symbol: Nsf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008740.4 | NP_032766.2 | 11 - 12 | 1259 | 64 | |
| GenBank mRNA | AK049281.1 | - | 11 - 12 | 1229 | 64 | |
| AK085086.1 | - | 11 - 12 | 1213 | 64 | ||
| AK153905.1 | - | 11 - 12 | 1228 | 64 | ||
| BC006627.1 | - | 11 - 12 | 1264 | 3968733 | 64 | |
| BC019167.1 | - | 11 - 12 | 1212 | 4506351 | 64 | |
| U10120.1 | - | 11 - 12 | 1217 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nsf
Genomic Map