Assay Details
| Gene Symbol: Nsf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008740.4 | NP_032766.2 | 15 - 16 | 1839 | 78 | |
| GenBank mRNA | AK028415.1 | - | 4 - 5 | 563 | 78 | |
| AK032120.1 | - | 2 - 3 | 271 | 78 | ||
| AK049281.1 | - | 15 - 16 | 1809 | 78 | ||
| AK085086.1 | - | 15 - 16 | 1793 | 78 | ||
| AK153905.1 | - | 15 - 16 | 1808 | 78 | ||
| AK181330.1 | - | 1 - 2 | 124 | 78 | ||
| BC006627.1 | - | 15 - 16 | 1844 | 3968733 | 78 | |
| BC019167.1 | - | 15 - 16 | 1792 | 4506351 | 78 | |
| U10120.1 | - | 15 - 16 | 1797 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nsf
Genomic Map