Assay Details
| Gene Symbol: Nsf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008740.4 | NP_032766.2 | 19 - 20 | 2233 | 104 | |
| GenBank mRNA | AK028415.1 | - | 8 - 9 | 957 | 104 | |
| AK032120.1 | - | 6 - 7 | 665 | 104 | ||
| AK049281.1 | - | 19 - 20 | 2203 | 104 | ||
| AK085086.1 | - | 19 - 20 | 2187 | 104 | ||
| AK153905.1 | - | 19 - 20 | 2202 | 104 | ||
| BC006627.1 | - | 19 - 20 | 2238 | 3968733 | 104 | |
| BC019167.1 | - | 19 - 20 | 2186 | 4506351 | 104 | |
| U10120.1 | - | 19 - 20 | 2191 | 104 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nsf
Genomic Map