Assay Details
| Gene Symbol: Fxyd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111073.2 | NP_001104543.1 | 1 - 2 | 445 | 85 | |
| NM_001287213.1 | NP_001274142.1 | 2 - 3 | 317 | 85 | ||
| NM_001287217.1 | NP_001274146.1 | 2 - 3 | 305 | 85 | ||
| NM_008761.4 | NP_032787.1 | 2 - 3 | 305 | 85 | ||
| GenBank mRNA | AI121044.1 | - | 2 - 3 | 305 | 85 | |
| AK003068.1 | - | 2 - 3 | 88 | 85 | ||
| AK155763.1 | - | 1 - 2 | 425 | 85 | ||
| AK169966.1 | - | 2 - 3 | 88 | 85 | ||
| AK170810.1 | - | 2 - 3 | 93 | 85 | ||
| BC013340.1 | - | 2 - 3 | 91 | 85 | ||
| BC031112.1 | - | 2 - 3 | 74 | 85 | ||
| CA528582.1 | - | 2 - 3 | 296 | 85 | ||
| U72680.1 | - | 2 - 3 | 111 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fxyd5
Genomic Map