Assay Details
| Gene Symbol: Tnfrsf11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008764.3 | NP_032790.3 | 2 - 3 | 638 | 119 | |
| GenBank mRNA | AB013898.1 | - | 2 - 3 | 119 | ||
| AK140244.1 | - | 2 - 3 | 461 | 119 | ||
| AK146105.1 | - | 2 - 3 | 581 | 119 | ||
| BC049782.1 | - | 2 - 3 | 6506550 | 119 | ||
| U94331.1 | - | 2 - 3 | 491 | 119 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tnfrsf11b
Genomic Map