Assay Details
| Gene Symbol: Pfkl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008826.4 | NP_032852.2 | 7 - 8 | 860 | 81 | |
| GenBank mRNA | AK030511.1 | - | 7 - 8 | 815 | 81 | |
| AK036318.1 | - | 7 - 8 | 793 | 81 | ||
| AK081313.1 | - | 7 - 8 | 811 | 81 | ||
| AK159328.1 | - | 7 - 8 | 805 | 81 | ||
| AK168660.1 | - | 7 - 8 | 816 | 81 | ||
| AK183616.1 | - | 1 - 2 | 79 | 81 | ||
| BC020097.1 | - | 7 - 8 | 814 | 4014395 | 81 | |
| BC029336.1 | - | 7 - 8 | 804 | 81 | ||
| J03928.1 | - | 7 - 8 | 789 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pfkl
Genomic Map