Assay Details
| Gene Symbol: Pfkl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008826.4 | NP_032852.2 | 15 - 16 | 1614 | 93 | |
| GenBank mRNA | AK030511.1 | - | 15 - 16 | 1569 | 93 | |
| AK030892.1 | - | 8 - 9 | 713 | 93 | ||
| AK036318.1 | - | 15 - 16 | 1547 | 93 | ||
| AK081313.1 | - | 15 - 16 | 1565 | 93 | ||
| AK159328.1 | - | 15 - 16 | 1559 | 93 | ||
| AK168660.1 | - | 15 - 16 | 1570 | 93 | ||
| AK208650.1 | - | 1 - 2 | 49 | 93 | ||
| BC020097.1 | - | 15 - 16 | 1568 | 4014395 | 93 | |
| BC029336.1 | - | 15 - 16 | 1557 | 93 | ||
| J03928.1 | - | 15 - 16 | 1543 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pfkl
Genomic Map