Assay Details
| Gene Symbol: Pfkl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008826.4 | NP_032852.2 | 19 - 20 | 2104 | 76 | |
| GenBank mRNA | AK030511.1 | - | 19 - 20 | 2059 | 76 | |
| AK030892.1 | - | 12 - 13 | 1203 | 76 | ||
| AK036318.1 | - | 19 - 20 | 2037 | 76 | ||
| AK081313.1 | - | 19 - 20 | 2055 | 76 | ||
| AK159328.1 | - | 19 - 20 | 2049 | 76 | ||
| AK168660.1 | - | 19 - 20 | 2060 | 76 | ||
| AK216193.1 | - | 3 - 4 | 213 | 76 | ||
| BC029336.1 | - | 19 - 20 | 2047 | 76 | ||
| J03928.1 | - | 19 - 20 | 2033 | 76 | ||
More Information
Set Membership: |
|

Pfkl
Genomic Map