Assay Details
| Gene Symbol: Pgr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008829.2 | NP_032855.2 | 5 - 6 | 2973 | 95 | |
| GenBank mRNA | AK054117.1 | - | 5 - 6 | 1654 | 95 | |
| BC145807.1 | - | 5 - 6 | 2378 | 40131110 | 95 | |
| M68915.1 | - | 5 - 6 | 2336 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pgr
Genomic Map