Assay Details
| Gene Symbol: Pik3ca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008839.2 | NP_032865.2 | 18 - 19 | 2843 | 102 | |
| GenBank mRNA | AK051885.1 | - | 18 - 19 | 2476 | 102 | |
| BC089038.1 | - | 18 - 19 | 2843 | 30692343 | 102 | |
| BC130228.1 | - | 18 - 19 | 2792 | 40141870 | 102 | |
| U03279.1 | - | 18 - 19 | 2788 | 102 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pik3ca
Genomic Map