Assay Details
| Gene Symbol: Pik3r2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008841.2 | NP_032867.2 | 7 - 8 | 1368 | 64 | |
| GenBank mRNA | AK146195.1 | - | 7 - 8 | 1358 | 64 | |
| AK148122.1 | - | 7 - 8 | 1365 | 64 | ||
| AK216667.1 | - | 3 | 303 | 65 | ||
| BC006796.1 | - | 7 - 8 | 1359 | 2581969 | 64 | |
| BC085501.1 | - | 6 - 7 | 1291 | 6816009 | 64 | |
| Y13569.1 | - | 7 - 8 | 1337 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pik3r2
Genomic Map