Assay Details
| Gene Symbol: Pik3r2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008841.2 | NP_032867.2 | 10 - 11 | 1758 | 71 | |
| GenBank mRNA | AK146195.1 | - | 10 - 11 | 1748 | 71 | |
| AK148122.1 | - | 10 - 11 | 1755 | 71 | ||
| AK184221.1 | - | 1 - 2 | 149 | 71 | ||
| BC006796.1 | - | 10 - 11 | 1749 | 2581969 | 71 | |
| BC085501.1 | - | 9 - 10 | 1681 | 6816009 | 71 | |
| Y13569.1 | - | 10 - 11 | 1727 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pik3r2
Genomic Map