Assay Details
| Gene Symbol: Pik3r2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008841.2 | NP_032867.2 | 13 - 14 | 2206 | 56 | |
| GenBank mRNA | AK146195.1 | - | 13 - 14 | 2196 | 56 | |
| AK148122.1 | - | 13 - 14 | 2203 | 56 | ||
| BC006796.1 | - | 13 - 14 | 2197 | 2581969 | 56 | |
| BC085501.1 | - | 12 - 13 | 2129 | 6816009 | 56 | |
| Y13569.1 | - | 13 - 14 | 2175 | 56 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pik3r2
Genomic Map