Assay Details
| Gene Symbol: Prkcb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008855.2 | NP_032881.1 | 10 - 11 | 1460 | 66 | |
| GenBank mRNA | AK051862.1 | - | 10 - 11 | 1453 | 66 | |
| BC127083.1 | - | 10 - 11 | 1279 | 40111744 | 66 | |
| X53532.1 | - | 10 - 11 | 1288 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prkcb
Genomic Map