Assay Details
| Gene Symbol: Prkch | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008856.3 | NP_032882.2 | 2 - 3 | 547 | 103 | |
| GenBank mRNA | AK076352.1 | - | 2 - 3 | 746 | 103 | |
| AK145817.1 | - | 2 - 3 | 793 | 103 | ||
| AK164044.1 | - | 2 - 3 | 746 | 103 | ||
| BC031121.1 | - | 2 - 3 | 547 | 5059633 | 103 | |
| D90242.1 | - | 2 - 3 | 484 | 103 | ||
More Information
Set Membership: |
|

Prkch
Genomic Map