Assay Details
| Gene Symbol: Prkch | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008856.3 | NP_032882.2 | 8 - 9 | 1229 | 92 | |
| GenBank mRNA | AK145817.1 | - | 8 - 9 | 1475 | 92 | |
| AK155951.1 | - | 9 - 10 | 1572 | 92 | ||
| AK164044.1 | - | 8 - 9 | 1428 | 92 | ||
| BC031121.1 | - | 8 - 9 | 1229 | 5059633 | 92 | |
| D90242.1 | - | 8 - 9 | 1166 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prkch
Genomic Map