Assay Details
| Gene Symbol: Prkcz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039079.2 | NP_001034168.1 | 5 - 6 | 692 | 124 | |
| NM_008860.3 | NP_032886.2 | 8 - 9 | 892 | 124 | ||
| GenBank mRNA | AB110830.1 | - | 5 - 6 | 652 | 124 | |
| AK042562.1 | - | 5 - 6 | 650 | 124 | ||
| AK049959.1 | - | 5 - 6 | 676 | 124 | ||
| AK147300.1 | - | 5 - 6 | 510 | 124 | ||
| BC072590.1 | - | 5 - 6 | 692 | 124 | ||
| BC139761.1 | - | 8 - 9 | 709 | 40109623 | 124 | |
| M94632.1 | - | 8 - 9 | 709 | 124 | ||
More Information
Set Membership: |
|

Prkcz
Genomic Map