Assay Details
| Gene Symbol: Prkcz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039079.2 | NP_001034168.1 | 10 - 11 | 1288 | 109 | |
| NM_008860.3 | NP_032886.2 | 13 - 14 | 1488 | 109 | ||
| GenBank mRNA | AB110830.1 | - | 10 - 11 | 1248 | 109 | |
| AK042562.1 | - | 10 - 11 | 1246 | 109 | ||
| AK049959.1 | - | 10 - 11 | 1272 | 109 | ||
| AK147300.1 | - | 10 - 11 | 1106 | 109 | ||
| BC057694.1 | - | 5 - 6 | 455 | 109 | ||
| BC072590.1 | - | 10 - 11 | 1288 | 109 | ||
| BC139761.1 | - | 13 - 14 | 1305 | 40109623 | 109 | |
| M94632.1 | - | 13 - 14 | 1305 | 109 | ||
More Information
Set Membership: |
|

Prkcz
Genomic Map