Assay Details
| Gene Symbol: Prl3b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008865.3 | NP_032891.1 | 1 - 2 | 89 | 71 | |
| GenBank mRNA | AK005542.1 | - | 1 - 2 | 90 | 71 | |
| AK005543.1 | - | 1 - 2 | 89 | 71 | ||
| AK131667.1 | - | 1 - 2 | 90 | 71 | ||
| AK167483.1 | - | 1 - 2 | 90 | 71 | ||
| BC099386.1 | - | 1 - 2 | 74 | 30787415 | 71 | |
| M14647.1 | - | 1 - 2 | 89 | 71 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK005542.1,AK005543.1,AK131667.1,AK167483.1,BC099386.1,M14647.1,NM_008865.3 transcripts.
Set Membership: |
|

Prl3b1
Genomic Map