Assay Details
| Gene Symbol: Prl3b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008865.3 | NP_032891.1 | 2 - 3 | 253 | 76 | |
| GenBank mRNA | AK005542.1 | - | 2 - 3 | 254 | 76 | |
| AK005543.1 | - | 2 - 3 | 253 | 76 | ||
| AK131667.1 | - | 2 - 3 | 254 | 76 | ||
| AK167483.1 | - | 2 - 3 | 254 | 76 | ||
| BC099386.1 | - | 2 - 3 | 238 | 30787415 | 76 | |
| M14647.1 | - | 2 - 3 | 253 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prl3b1
Genomic Map