Assay Details
| Gene Symbol: Prkar1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253890.1 | NP_001240819.1 | 8 - 9 | 899 | 70 | |
| NM_008923.3 | NP_032949.3 | 8 - 9 | 901 | 70 | ||
| GenBank mRNA | AK161621.1 | - | 8 - 9 | 832 | 70 | |
| BC011424.1 | - | 8 - 9 | 840 | 3674751 | 70 | |
| CO425808.1 | - | 4 - 5 | 314 | 70 | ||
| M20473.1 | - | 8 - 9 | 847 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prkar1b
Genomic Map