Assay Details
| Gene Symbol: Prkar2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008924.2 | NP_032950.1 | 2 - 3 | 605 | 105 | |
| GenBank mRNA | AF533977.1 | - | 2 - 3 | 605 | 105 | |
| AK132728.1 | - | 2 - 3 | 544 | 105 | ||
| BC075623.1 | - | 2 - 3 | 576 | 6854057 | 105 | |
| BC080276.1 | - | 2 - 3 | 599 | 30354403 | 105 | |
| J02935.1 | - | 2 - 3 | 288 | 105 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prkar2a
Genomic Map