Assay Details
| Gene Symbol: Tmprss15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008941.3 | NP_032967.1 | 6 - 7 | 759 | 67 | |
| NM_178855.4 | NP_849186.2 | 6 - 7 | 759 | 67 | ||
| GenBank mRNA | AK033582.1 | - | 6 - 7 | 761 | 67 | |
| BC117917.1 | - | 6 - 7 | 758 | 40097367 | 67 | |
| BC117918.1 | - | 6 - 7 | 758 | 40097370 | 67 | |
| U73378.1 | - | 6 - 7 | 754 | 67 | ||
More Information
Set Membership: |
|

Tmprss15
Genomic Map