Assay Details
| Gene Symbol: Ptgdr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008962.4 | NP_032988.3 | 1 - 2 | 963 | 86 | |
| GenBank mRNA | AK032488.1 | - | 1 - 2 | 964 | 86 | |
| BC132297.1 | - | 1 - 2 | 942 | 40130574 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
inflammatory response
signal transduction
G-protein coupled receptor signaling pathway
male sex determination
sleep
adenosine metabolic process
cellular response to prostaglandin D stimulus
prostaglandin J receptor activity
signal transducer activity
G-protein coupled receptor activity
prostaglandin D receptor activity
inflammatory response
signal transduction
G-protein coupled receptor signaling pathway
male sex determination
sleep
adenosine metabolic process
cellular response to prostaglandin D stimulus
prostaglandin J receptor activity
signal transducer activity
G-protein coupled receptor activity
prostaglandin D receptor activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29567894 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29567893 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29567891 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2148204 | NM_008962.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2148204 | AK032488.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 218011 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 455002 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 449464 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 829777 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 834334 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 24342 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P112761 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2148204 | NM_008962.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2148204 | AK032488.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P36889 | BC132297.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P36889 | NM_008962.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P36889 | AK032488.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P292354 | BC132297.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P292354 | NM_008962.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P292354 | AK032488.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29567923 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29567900 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29567908 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29567911 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29567928 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29567909 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29567887 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29567930 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29567914 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29567897 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29567919 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29567920 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29567921 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29567913 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29567895 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29567889 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29567894 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29567917 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29567883 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29567903 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29567925 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29567906 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29567891 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29567927 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29567901 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29567896 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29567885 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29567904 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29567916 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29567912 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29567893 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29567881 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P112761 | - | - |
| Whole Mouse Genome G4122A | A_52_P36889 | BC132297.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P36889 | NM_008962.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P36889 | AK032488.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P292354 | BC132297.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P292354 | NM_008962.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P292354 | AK032488.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P105017 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139011 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P112761 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29567891 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29567893 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29567894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29567803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29567848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29567918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29567852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29567925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29567877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29567822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29567793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29567816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29567921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29567894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29567887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29567832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29567914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29567917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29567791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29567851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29567885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29567901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29567795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29567867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29567938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29567812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29567891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29567883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29567946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29567947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29567801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29567912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29567815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29567846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29567797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29567798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29567806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29567908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29567911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29567854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29567928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29567939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29567788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29567808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29567923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29567913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29567903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29567941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29567840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29567906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29567945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29567845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29567896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29567785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29567934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29567942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29567783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29567804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29567843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29567933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29567889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29567895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29567930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29567787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29567874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29567949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29567796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29567881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29567893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29567799 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29567848 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29567916 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29567877 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29567938 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29567881 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29567918 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29567934 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29567845 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29567859 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29567840 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29567852 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29567880 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29567908 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29567843 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29567920 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29567854 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29567917 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29567930 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29567904 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29567895 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29567933 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29567928 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29567860 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29567912 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29567942 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29567875 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29567911 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29567914 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29567936 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29567923 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29567939 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29567874 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29567855 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29567891 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29567941 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29567921 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29567903 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29567879 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29567893 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29567943 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29567887 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29567906 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29567935 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29567885 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29567853 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29567846 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29567851 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29567919 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29567925 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29567883 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29567913 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29567901 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29567896 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29567889 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29567894 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29567867 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29567897 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10419169 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10419168 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422171_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10419167 | - | - |
| Mouse Genome 430 2.0 Array | 1422171_at | - | - |
| Mouse Genome 430 2.0 Array | 1422171_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 19214_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 19214_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1400014263.mm.1 | NM_008962.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400014263.mm.1 | NM_008962.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400014263.mm.1 | TC1400001892.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1400014264.mm.1 | NM_008962.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400014264.mm.1 | NM_008962.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400014264.mm.1 | TC1400001892.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1400014262.mm.1 | NM_008962.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400014262.mm.1 | NM_008962.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400014262.mm.1 | TC1400001892.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1422171_at | - | - |
| Mouse Expression Set 430 | 1422171_PM_at | - | - |
| Murine Genome U74v2 Set | 98856_at | - | - |
| Murine Genome U74v2 Set | 98856_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422171_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422171_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422171_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422171_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 533506 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 3850161 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003850161 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1247634 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1247634 | - | - |
| mouse-6 v1.1 (Illumina) | scl45678.2.1_305-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3850161 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl45678.2.1_305-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3850161 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1247634 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1247634 | - | - |

Ptgdr
Genomic Map