Assay Details
| Gene Symbol: Abcd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008992.2 | NP_033018.2 | 4 - 5 | 568 | 52 | |
| GenBank mRNA | AJ001166.1 | - | 4 - 5 | 472 | 52 | |
| AK028420.1 | - | 4 - 5 | 473 | 52 | ||
| BC050102.1 | - | 4 - 5 | 676 | 30012556 | 52 | |
More Information
Updated Mapping Information
Set Membership: |
|

Abcd4
Genomic Map