Assay Details
| Gene Symbol: Abcd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008992.2 | NP_033018.2 | 17 - 18 | 1783 | 60 | |
| GenBank mRNA | AJ001166.1 | - | 17 - 18 | 1687 | 60 | |
| AK028420.1 | - | 17 - 18 | 1840 | 60 | ||
More Information
Set Membership: |
|

Abcd4
Genomic Map