Assay Details
| Gene Symbol: Rab4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009003.3 | NP_033029.2 | 1 - 2 | 168 | 79 | |
| GenBank mRNA | AK136566.1 | - | 1 - 2 | 157 | 79 | |
| AK145452.1 | - | 1 - 2 | 151 | 79 | ||
| AK161877.1 | - | 1 - 2 | 136 | 79 | ||
| BC138508.1 | - | 1 - 2 | 81 | 8861528 | 79 | |
| CO806247.1 | - | 184 | 79 | |||
| D86563.1 | - | 1 - 2 | 142 | 79 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK136566.1,AK145452.1,AK161877.1,BC138508.1,D86563.1,NM_009003.3 transcripts.
Set Membership: |
|

Rab4a
Genomic Map