Assay Details
| Gene Symbol: Rara | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001176528.1 | NP_001169999.1 | 3 - 4 | 955 | 72 | |
| NM_001177302.1 | NP_001170773.1 | 5 - 6 | 1160 | 72 | ||
| NM_001177303.1 | NP_001170774.1 | 4 - 5 | 905 | 72 | ||
| NM_009024.2 | NP_033050.2 | 4 - 5 | 1027 | 72 | ||
| GenBank mRNA | AB221572.1 | - | 3 - 4 | 474 | 72 | |
| AK042163.1 | - | 4 - 5 | 906 | 72 | ||
| AK153644.1 | - | 3 - 4 | 501 | 72 | ||
| AK154623.1 | - | 4 - 5 | 1000 | 72 | ||
| AK170956.1 | - | 3 - 4 | 955 | 72 | ||
| BC010216.1 | - | 4 - 5 | 1025 | 3498142 | 72 | |
| BC038266.1 | - | 3 - 4 | 1024 | 72 | ||
| BC040383.1 | - | 5 - 6 | 1160 | 72 | ||
| CT010231.1 | - | 3 - 4 | 470 | 72 | ||
| S56656.1 | - | 4 - 5 | 1007 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rara
Genomic Map