Assay Details
| Gene Symbol: Ret | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080780.1 | NP_001074249.1 | 13 - 14 | 3009 | 82 | |
| NM_009050.2 | NP_033076.2 | 13 - 14 | 3009 | 82 | ||
| GenBank mRNA | AF209436.1 | - | 13 - 14 | 2398 | 82 | |
| AK044686.1 | - | 13 - 14 | 2596 | 82 | ||
| AK051633.1 | - | 13 - 14 | 2588 | 82 | ||
| AK084977.1 | - | 13 - 14 | 2659 | 82 | ||
| AY326397.1 | - | 13 - 14 | 2398 | 82 | ||
| BC059012.1 | - | 13 - 14 | 2983 | 6849696 | 82 | |
| X67812.1 | - | 13 - 14 | 2398 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ret
Genomic Map