| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0600023014.mm.1 |
TC0600002996.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600023014.mm.1 |
NM_001080780.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600023014.mm.1 |
NM_001080780.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600023014.mm.1 |
NM_009050.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600023014.mm.1 |
NM_009050.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600023013.mm.1 |
TC0600002996.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600023013.mm.1 |
NM_001080780.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600023013.mm.1 |
NM_001080780.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600023013.mm.1 |
NM_009050.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600023013.mm.1 |
NM_009050.2 |
Refseq |