Assay Details
| Gene Symbol: Rps6ka1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285505.1 | NP_001272434.1 | 19 - 20 | 1961 | 58 | |
| NM_001285506.1 | NP_001272435.1 | 18 - 19 | 1845 | 58 | ||
| NM_009097.5 | NP_033123.2 | 19 - 20 | 1994 | 58 | ||
| GenBank mRNA | AK132856.1 | - | 17 - 18 | 2031 | 58 | |
| AK160571.1 | - | 7 - 8 | 789 | 58 | ||
| AK167790.1 | - | 18 - 19 | 1832 | 58 | ||
| BC049076.1 | - | 19 - 20 | 1945 | 58 | ||
| BC094470.1 | - | 19 - 20 | 1947 | 5098650 | 58 | |
| M28489.1 | - | 19 - 20 | 1947 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rps6ka1
Genomic Map