Assay Details
| Gene Symbol: Rrh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009102.3 | NP_033128.1 | 1 - 2 | 125 | 65 | |
| GenBank mRNA | AF012271.1 | - | 1 - 2 | 129 | 65 | |
| AK044717.1 | - | 113 | 65 | |||
| BC046288.1 | - | 1 - 2 | 125 | 6468540 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rrh
Genomic Map