Assay Details
| Gene Symbol: Rxrg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159731.1 | NP_001153203.1 | 7 - 8 | 1071 | 75 | |
| NM_009107.3 | NP_033133.1 | 8 - 9 | 1527 | 75 | ||
| GenBank mRNA | AK132607.1 | - | 1 - 2 | 95 | 75 | |
| BC058401.1 | - | 8 - 9 | 1509 | 5707723 | 75 | |
| G49237.1 | - | 8 - 9 | 1259 | 75 | ||
| M84819.1 | - | 8 - 9 | 1172 | 75 | ||
| S62948.1 | - | 8 - 9 | 1371 | 75 | ||
| S62952.1 | - | 7 - 8 | 1027 | 75 | ||
| X66225.1 | - | 8 - 9 | 1259 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rxrg
Genomic Map