Assay Details
| Gene Symbol: Nr1h4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163504.1 | NP_001156976.1 | 3 - 4 | 726 | 60 | |
| NM_001163700.1 | NP_001157172.1 | 2 - 3 | 522 | 60 | ||
| NM_009108.2 | NP_033134.2 | 2 - 3 | 522 | 60 | ||
| GenBank mRNA | AK002513.1 | - | 2 - 3 | 508 | 60 | |
| AK132255.1 | - | 2 - 3 | 514 | 60 | ||
| BC015261.1 | - | 2 - 3 | 493 | 4241705 | 60 | |
| U09416.1 | - | 2 - 3 | 491 | 60 | ||
| U09417.1 | - | 3 - 4 | 690 | 60 | ||
| U09418.1 | - | 3 - 4 | 687 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Citations (1)
Fibroblast growth factor 21 corrects obesity in mice.
- Authors
- Coskun T, Bina HA, Schneider MA, Dunbar JD, Hu CC, Chen Y, Moller DE, Kharitonenkov A
- Journal
- Endocrinology - Page No 6018 (Mon Dec 01 00:00:00 EST 2008)
- Volume (Year)
- 149 (2008)
- Product Usage
- -

Nr1h4
Genomic Map