Assay Details
| Gene Symbol: Nr1h4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009108.2 | NP_033134.2 | 3 - 4 | 654 | 108 | |
| GenBank mRNA | AK132255.1 | - | 3 - 4 | 646 | 108 | |
| BC015261.1 | - | 3 - 4 | 625 | 4241705 | 108 | |
| U09416.1 | - | 3 - 4 | 623 | 108 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nr1h4
Genomic Map