Assay Details
| Gene Symbol: Sema3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042779.2 | NP_001036244.1 | 11 - 12 | 1320 | 68 | |
| NM_001291537.1 | NP_001278466.1 | 11 - 12 | 1584 | 68 | ||
| NM_001291538.1 | NP_001278467.1 | 11 - 12 | 1578 | 68 | ||
| NM_001291539.1 | NP_001278468.1 | 10 - 11 | 1261 | 68 | ||
| NM_009153.3 | NP_033179.2 | 9 - 10 | 1130 | 68 | ||
| NR_111986.1 | 9 - 10 | 1140 | 68 | |||
| GenBank mRNA | AK014333.1 | - | 9 - 10 | 1076 | 68 | |
| AK033597.1 | - | 10 - 11 | 1271 | 68 | ||
| AK037034.1 | - | 11 - 12 | 1309 | 68 | ||
| AK043634.1 | - | 10 - 11 | 1214 | 68 | ||
| AK048364.1 | - | 10 - 11 | 1542 | 68 | ||
| BC090669.1 | - | 10 - 11 | 1241 | 30696359 | 68 | |
| BC150758.1 | - | 8 - 9 | 976 | 9087669 | 68 | |
| CN538654.1 | - | 2 - 3 | 176 | 68 | ||
| X85990.1 | - | 9 - 10 | 1127 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sema3b
Genomic Map