Assay Details
| Gene Symbol: Slc2a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009204.2 | NP_033230.2 | 6 - 7 | 934 | 88 | |
| GenBank mRNA | AB008453.1 | - | 6 - 7 | 732 | 88 | |
| AK137607.1 | - | 6 - 7 | 919 | 88 | ||
| AK138832.1 | - | 6 - 7 | 908 | 88 | ||
| AK160882.1 | - | 5 - 6 | 1035 | 88 | ||
| AK164310.1 | - | 6 - 7 | 909 | 88 | ||
| BC014282.1 | - | 6 - 7 | 904 | 4207674 | 88 | |
| M23383.1 | - | 6 - 7 | 88 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Slc2a4
Genomic Map